Anti-IL3 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:500 to 1:1000
Type:
Antigen: IL3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77809-993
- 77809-994
- Antigen symbol:
- IL3
- IL3
- Antigen name:
- Interleukin-3 protein (20-152AA)
- Interleukin-3 protein (20-152AA)
- Antigen synonyms:
- Colony stimulating factor multiple| Hematopoietic growth factor| IL 3| IL-3| IL3| IL3_HUMAN| Interleukin 3 (colony stimulating factor| multiple)| Interleukin 3| Interleukin-3| Mast cell growth factor| MCGF| MGC79398| MGC79399| Multi CSF| Multilineage colony stimulating factor| Multipotential colony stimulating factor| Multipotential colony-stimulating factor| OTTHUMP00000065963| P cell stimulating factor | P-cell-stimulating factor
- Colony stimulating factor multiple| Hematopoietic growth factor| IL 3| IL-3| IL3| IL3_HUMAN| Interleukin 3 (colony stimulating factor| multiple)| Interleukin 3| Interleukin-3| Mast cell growth factor| MCGF| MGC79398| MGC79399| Multi CSF| Multilineage colony stimulating factor| Multipotential colony stimulating factor| Multipotential colony-stimulating factor| OTTHUMP00000065963| P cell stimulating factor | P-cell-stimulating factor
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P08700
- P08700
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together