Anti-NPY6R Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NPY6R
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77834-393
- 77834-394
- Antigen symbol:
- NPY6R
- NPY6R
- Antigen name:
- Putative neuropeptide Y receptor type 6 protein (1-39AA)
- Putative neuropeptide Y receptor type 6 protein (1-39AA)
- Antigen synonyms:
- NPY6R|NPY1RL|Y2B|Putative neuropeptide Y receptor type 6|NPY6-R|NPY Y1-like receptor|Putative pancreatic polypeptide receptor 2|PP2
- NPY6R|NPY1RL|Y2B|Putative neuropeptide Y receptor type 6|NPY6-R|NPY Y1-like receptor|Putative pancreatic polypeptide receptor 2|PP2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99463
- Q99463
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together