Anti-NIF3L1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NIF3L1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-498
- 77840-499
- Antigen symbol:
- NIF3L1
- NIF3L1
- Antigen name:
- NIF3-like protein 1 protein (1-377AA)
- NIF3-like protein 1 protein (1-377AA)
- Antigen synonyms:
- ALS2CR1| Amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 1| Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 1 protein| CALS-7| MDS015| Ngg1 interacting factor 3-like 1| NIF3 (Ngg1 interacting factor 3, S.pombe homolog)-like 1| NIF3 NGG1 interacting factor 3-like 1 (S. pombe)| NIF3 NGG1 interacting factor 3-like 1| NIF3-like protein 1| NIF3L_HUMAN| Nif3l1| OTTHUMP00000206190| OTTHUMP00000206191| OTTHUMP00000206192
- ALS2CR1| Amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 1| Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 1 protein| CALS-7| MDS015| Ngg1 interacting factor 3-like 1| NIF3 (Ngg1 interacting factor 3, S.pombe homolog)-like 1| NIF3 NGG1 interacting factor 3-like 1 (S. pombe)| NIF3 NGG1 interacting factor 3-like 1| NIF3-like protein 1| NIF3L_HUMAN| Nif3l1| OTTHUMP00000206190| OTTHUMP00000206191| OTTHUMP00000206192
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9GZT8
- Q9GZT8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together