Anti-KIR2DS5 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: KIR2DS5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-997
- 77823-996
- Antigen symbol:
- KIR2DS5
- KIR2DS5
- Antigen name:
- Killer cell immunoglobulin-like receptor 2DS5 protein (162-242AA)
- Killer cell immunoglobulin-like receptor 2DS5 protein (162-242AA)
- Antigen synonyms:
- KIR2DS5|CD158G|NKAT9Killer cell immunoglobulin-like receptor 2DS5|CD158 antigen-like family member G|MHC class I NK cell receptor|Natural killer-associated transcript 9|NKAT-9|CD antigen CD158g
- KIR2DS5|CD158G|NKAT9Killer cell immunoglobulin-like receptor 2DS5|CD158 antigen-like family member G|MHC class I NK cell receptor|Natural killer-associated transcript 9|NKAT-9|CD antigen CD158g
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14953
- Q14953
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together