Anti-PHKG1 Rabbit Polyclonal Antibody
77807-874
New Product
- Antigen name:Synthesized peptide derived from internal of PHKG1.
- Antigen symbol:PHKG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 μl
- Form:Liquid
- Gene ID:Q16816
- Antigen synonyms:PHK gamma M|PHK-gamma-M|Phkg|PHKG1_HUMAN|PHKIN01|Phosphorylase b kinase gamma catalytic chain|Phosphorylase b kinase gamma catalytic chain skeletal muscle/heart isoform|phosphorylase b kinase gamma catalytic chain, skeletal muscle isoform|phosphorylase kinase gamma|phosphorylase kinase subunit gamma 1|Phosphorylase kinase subunit gamma-1|phosphorylase kinase, gamma 1 (muscle)|Phosphorylase kinase, muscle, gamma-1|Serine/threonine-protein kinase PHKG1|skeletal muscle/heart isoform
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on Blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: PHKG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human