Anti-APOLD1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: APOLD1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77833-101
- 77833-102
- Antigen symbol:
- APOLD1
- APOLD1
- Antigen name:
- Apolipoprotein L domain-containing protein 1 protein (213-274AA)
- Apolipoprotein L domain-containing protein 1 protein (213-274AA)
- Antigen synonyms:
- APLD1_HUMAN|APOLD1|Apolipoprotein L domain containing protein 1|Apolipoprotein L domain-containing protein 1|Vascular early response gene protein|VERGE
- APLD1_HUMAN|APOLD1|Apolipoprotein L domain containing protein 1|Apolipoprotein L domain-containing protein 1|Vascular early response gene protein|VERGE
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96LR9
- Q96LR9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together