Anti-PIP4P2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77803-086
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Type 2 phosphatidylinositol 4,5-bisphosphate 4-phosphatase protein (1-191AA)
- Antigen symbol:PIP4P2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q8N4L2
- Antigen synonyms:2610319K07Rik|5-bisphosphate 4-phosphatase|5-P2 4-Ptase II|AI315591|AV001360|PtdIns-4|RGD1306225|RP23-379L17.2|TM55A_HUMAN|tmem55a|Transmembrane protein 55A|Type II phosphatidylinositol 4
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: PIP4P2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse