Anti-ATP5ME Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ATP5ME
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77808-709
- 77808-708
- Antigen symbol:
- ATP5ME
- ATP5ME
- Antigen name:
- ATP synthase subunit e, mitochondrial protein (2-69AA)
- ATP synthase subunit e, mitochondrial protein (2-69AA)
- Antigen synonyms:
- ATP 5I|ATP 5K|ATP synthase e chain mitochondrial|ATP synthase H+ transporting mitochondrial F0 complex subunit E|ATP synthase subunit e|ATP synthase subunit e mitochondrial|ATP5I|ATP5I_HUMAN|ATP5K|ATPase subunit e|F1F0 ATP synthase murine e subunit|MGC12532|mitochondrial
- ATP 5I|ATP 5K|ATP synthase e chain mitochondrial|ATP synthase H+ transporting mitochondrial F0 complex subunit E|ATP synthase subunit e|ATP synthase subunit e mitochondrial|ATP5I|ATP5I_HUMAN|ATP5K|ATPase subunit e|F1F0 ATP synthase murine e subunit|MGC12532|mitochondrial
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P56385
- P56385
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together