Anti-MMP16 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: MMP16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77821-440
- 77821-439
- Antigen symbol:
- MMP16
- MMP16
- Antigen name:
- Fusion protein of MMP16
- Fusion protein of MMP16
- Antigen synonyms:
- C8orf57|DKFZp761D112|Matrix metalloproteinase 16|Matrix metalloproteinase-16|Membrane type 3 matrix metalloproteinase|Membrane type matrix metalloproteinase 3|Membrane-type matrix metalloproteinase 3|Membrane-type-3 matrix metalloproteinase|MMP 16|MMP-16|MMP-X2|MMP16_HUMAN|MT-MMP 3|MT-MMP2|MT-MMP3|MT3-MMP|MT3MMP|MTMMP2|MTMMP3|Putative transmembrane protein C8orf57
- C8orf57|DKFZp761D112|Matrix metalloproteinase 16|Matrix metalloproteinase-16|Membrane type 3 matrix metalloproteinase|Membrane type matrix metalloproteinase 3|Membrane-type matrix metalloproteinase 3|Membrane-type-3 matrix metalloproteinase|MMP 16|MMP-16|MMP-X2|MMP16_HUMAN|MT-MMP 3|MT-MMP2|MT-MMP3|MT3-MMP|MT3MMP|MTMMP2|MTMMP3|Putative transmembrane protein C8orf57
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P51512
- P51512
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together