Anti-ZIP1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ZIP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Saccharomyces cerevisiae
Specifications
- Catalog No:
- 77816-234
- 77816-233
- Antigen symbol:
- ZIP1
- ZIP1
- Antigen name:
- Recombinant Saccharomyces cerevisiae Synaptonemal complex protein ZIP1 protein (1-227AA)
- Recombinant Saccharomyces cerevisiae Synaptonemal complex protein ZIP1 protein (1-227AA)
- Antigen synonyms:
- ZIP1|YDR285W|D9819.9Synaptonemal complex protein ZIP1
- ZIP1|YDR285W|D9819.9Synaptonemal complex protein ZIP1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- S. cerevisiae
- S. cerevisiae
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P31111
- P31111
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together