Anti-TPP2 Rabbit Polyclonal Antibody
77803-388
New Product
- Antigen name:TPP-II
- Antigen symbol:TPP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 μl
- Form:Liquid
- Gene ID:P29144
- Antigen synonyms:Tripeptidyl-peptidase 2|TPP-2|EC 3.4.14.10|Tripeptidyl aminopeptidase|Tripeptidyl-peptidase II
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on Blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: TPP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat