Anti-KCNC3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: KCNC3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77824-377
- 77824-376
- Antigen symbol:
- KCNC3
- KCNC3
- Antigen name:
- Potassium voltage-gated channel subfamily C member 3 protein (638-745AA)
- Potassium voltage-gated channel subfamily C member 3 protein (638-745AA)
- Antigen synonyms:
- Kcnc3|KCNC3_HUMAN|KSHIIID|KV3.3|Potassium voltage gated channel Shaw related subfamily member 3|Potassium voltage gated channel subfamily C member 3|Potassium voltage-gated channel subfamily C member 3|SCA13|Shaw related subfamily, member 3|Shaw related voltage gated potassium channel protein 3|Spinocerebellar ataxia 13|Voltage gated potassium channel protein KV3.3|Voltage gated potassium channel subunit Kv3.3|Voltage-gated potassium channel subunit Kv3.3
- Kcnc3|KCNC3_HUMAN|KSHIIID|KV3.3|Potassium voltage gated channel Shaw related subfamily member 3|Potassium voltage gated channel subfamily C member 3|Potassium voltage-gated channel subfamily C member 3|SCA13|Shaw related subfamily, member 3|Shaw related voltage gated potassium channel protein 3|Spinocerebellar ataxia 13|Voltage gated potassium channel protein KV3.3|Voltage gated potassium channel subunit Kv3.3|Voltage-gated potassium channel subunit Kv3.3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14003
- Q14003
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together