Anti-PTOV1 Rabbit Recombinant Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PTOV1
Clonality: Recombinant
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77832-043
- 77832-044
- Antigen symbol:
- PTOV1
- PTOV1
- Antigen name:
- Prostate tumor-overexpressed gene 1 protein (2-137AA)
- Prostate tumor-overexpressed gene 1 protein (2-137AA)
- Antigen synonyms:
- PTOV1|ACID2|PP642|UNQ6127/PRO20092|Prostate tumor-overexpressed gene 1 protein|PTOV-1|Activator interaction domain-containing protein 2
- PTOV1|ACID2|PP642|UNQ6127/PRO20092|Prostate tumor-overexpressed gene 1 protein|PTOV-1|Activator interaction domain-containing protein 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86YD1
- Q86YD1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together