Anti-SPTBN1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SPTBN1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77829-052
- 77829-051
- Antigen symbol:
- SPTBN1
- SPTBN1
- Antigen name:
- Spectrin beta chain, non-erythrocytic 1 protein (2096-2256AA)
- Spectrin beta chain, non-erythrocytic 1 protein (2096-2256AA)
- Antigen synonyms:
- Beta G spectrin|Beta II spectrin|Beta spectrin 2|Beta spectrin II|Beta-II spectrin|betaSpII|brain 1|ELF|Embryonic liver beta fodrin|Fodrin beta chain|non-erythroid beta chain 1|Spectrin|Spectrin beta chain|Spectrin, beta, non erythrocytic 1|Spectrin, non erythroid beta chain 1|SPTB2|SPTB2_HUMAN|SPTBN1
- Beta G spectrin|Beta II spectrin|Beta spectrin 2|Beta spectrin II|Beta-II spectrin|betaSpII|brain 1|ELF|Embryonic liver beta fodrin|Fodrin beta chain|non-erythroid beta chain 1|Spectrin|Spectrin beta chain|Spectrin, beta, non erythrocytic 1|Spectrin, non erythroid beta chain 1|SPTB2|SPTB2_HUMAN|SPTBN1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q01082
- Q01082
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together