Anti-SLC20A2 Rabbit Recombinant Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: SLC20A2
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77821-552
- 77821-551
- Antigen symbol:
- SLC20A2
- SLC20A2
- Antigen name:
- Sodium-dependent phosphate transporter 2 protein (235-485AA)
- Sodium-dependent phosphate transporter 2 protein (235-485AA)
- Antigen synonyms:
- Gibbon ape leukemia virus receptor 2|Gibbon leukemia virus receptor 2|GLVR 2|GLVR-2|GLVR2|hPit2|IBGC3|MLVAR|Murine leukemia virus amphotropic receptor|murine leukemia virus, amphotropic, receptor for|Phosphate transporter 2|PIT 2|PiT-2|Pit2|RAM1|S20A2_HUMAN|SLC20A2|Sodium-dependent phosphate transporter 2|Solute carrier family 20 (phosphate transporter) member 2|solute carrier family 20 (phosphate transporter), member 2|Solute carrier family 20 member 2
- Gibbon ape leukemia virus receptor 2|Gibbon leukemia virus receptor 2|GLVR 2|GLVR-2|GLVR2|hPit2|IBGC3|MLVAR|Murine leukemia virus amphotropic receptor|murine leukemia virus, amphotropic, receptor for|Phosphate transporter 2|PIT 2|PiT-2|Pit2|RAM1|S20A2_HUMAN|SLC20A2|Sodium-dependent phosphate transporter 2|Solute carrier family 20 (phosphate transporter) member 2|solute carrier family 20 (phosphate transporter), member 2|Solute carrier family 20 member 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q08357
- Q08357
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together