Anti-VPS51 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: VPS51
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77841-493
- 77841-494
- Antigen symbol:
- VPS51
- VPS51
- Antigen name:
- Vacuolar protein sorting-associated protein 51 homolog protein (595-673AA)
- Vacuolar protein sorting-associated protein 51 homolog protein (595-673AA)
- Antigen synonyms:
- ANG2|ANG3|Another new gene 2 protein|C11orf3|chromosome 11 open reading frame 2|Fat free homolog|ffr|FFR_HUMAN|FFR|FLJ13879|FLJ36472|Protein fat-free homolog|Vacuolar protein sorting 51 homolog|VPS51
- ANG2|ANG3|Another new gene 2 protein|C11orf3|chromosome 11 open reading frame 2|Fat free homolog|ffr|FFR_HUMAN|FFR|FLJ13879|FLJ36472|Protein fat-free homolog|Vacuolar protein sorting 51 homolog|VPS51
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UID3
- Q9UID3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together