Anti-RHOD Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RHOD
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77797-320
- 77797-321
- Antigen symbol:
- RHOD
- RHOD
- Antigen name:
- Rho-related GTP-binding protein RhoD protein (1-80AA)
- Rho-related GTP-binding protein RhoD protein (1-80AA)
- Antigen synonyms:
- ARHD|Ras homolog D|Ras homolog gene family member A|Ras homolog gene family member D|Rho|RHO D|Rho related GTP binding protein RhoD|Rho related protein HP1|Rho-related GTP-binding protein RhoD|Rho-related protein HP1|RHOD|RHOD_HUMAN|RhoHP1|RHOM
- ARHD|Ras homolog D|Ras homolog gene family member A|Ras homolog gene family member D|Rho|RHO D|Rho related GTP binding protein RhoD|Rho related protein HP1|Rho-related GTP-binding protein RhoD|Rho-related protein HP1|RHOD|RHOD_HUMAN|RhoHP1|RHOM
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O00212
- O00212
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together