Order Entry
Canada
ContactUsLinkComponent
Anti-TCIRG1 Rabbit Recombinant Polyclonal Antibody (HRP)
Anti-TCIRG1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-TCIRG1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77822-897
  • 77822-898
  • Antigen symbol:
  • TCIRG1
  • TCIRG1
  • Antigen name:
  • V-type proton ATPase 116 kDa subunit a isoform 3 protein (76-182AA)
  • V-type proton ATPase 116 kDa subunit a isoform 3 protein (76-182AA)
  • Antigen synonyms:
  • a3|Atp 6i|Atp6i|ATP6N1C|ATP6V0A3|ATPase H+ transporting 116kD|OC 116|OC 116 kDa|OC 116kDa|OC-116 kDa|OC116|OPTB 1|OPTB1|Osteoclastic proton pump 116 kDa subunit|Specific 116 kDa vacuolar proton pump subunit|Stv 1|Stv1|T cell immune regulator 1|T cell immune regulator 1 ATPase H+ transporting lysosomal V0 subunit A|T cell immune regulator 1 ATPase H+ transporting lysosomal V0 subunit A3|T cell immune response cDNA 7|T cell immune response cDNA7 protein|T cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein a|T-cell immune regulator 1|T-cell immune response cDNA7 protein|TCIRG 1|TCIRG1|TIRC 7|TIRC7|V ATPase 116 kDa|V ATPase 116 kDa isoform a3|V type proton ATPase 116 kDa subunit a|V-ATPase 116 kDa isoform a3|V-type proton ATPase 116 kDa subunit a isoform 3|Vacuolar proton translocating ATPase 116 kDa subunit A|Vacuolar proton translocating ATPase 116 kDa subunit a isoform 3|Vph 1|Vph1|VPP3_HUMAN
  • a3|Atp 6i|Atp6i|ATP6N1C|ATP6V0A3|ATPase H+ transporting 116kD|OC 116|OC 116 kDa|OC 116kDa|OC-116 kDa|OC116|OPTB 1|OPTB1|Osteoclastic proton pump 116 kDa subunit|Specific 116 kDa vacuolar proton pump subunit|Stv 1|Stv1|T cell immune regulator 1|T cell immune regulator 1 ATPase H+ transporting lysosomal V0 subunit A|T cell immune regulator 1 ATPase H+ transporting lysosomal V0 subunit A3|T cell immune response cDNA 7|T cell immune response cDNA7 protein|T cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein a|T-cell immune regulator 1|T-cell immune response cDNA7 protein|TCIRG 1|TCIRG1|TIRC 7|TIRC7|V ATPase 116 kDa|V ATPase 116 kDa isoform a3|V type proton ATPase 116 kDa subunit a|V-ATPase 116 kDa isoform a3|V-type proton ATPase 116 kDa subunit a isoform 3|Vacuolar proton translocating ATPase 116 kDa subunit A|Vacuolar proton translocating ATPase 116 kDa subunit a isoform 3|Vph 1|Vph1|VPP3_HUMAN
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • HRP (Horseradish Peroxidase)
  • HRP (Horseradish Peroxidase)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q13488
  • Q13488
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together