Anti-ABO Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: ABO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77807-937
- 77807-936
- Antigen symbol:
- ABO
- ABO
- Antigen name:
- Synthetic peptide of ABO
- Synthetic peptide of ABO
- Antigen synonyms:
- A transferase|A3GALNT|A3GALT1|ABO A3 transferase|Abo|ABO blood group (transferase A alpha 1-3 N acetylgalactosaminyltransferase transferase B alpha 1 3 galactosyltransferase)|ABO glycosyltransferase|ABO weak transfer|B (A) alpha 1|3 glactosyltransferase|B transferase|BGAT_HUMAN|Fucosylglycoprotein 3 alpha galactosyltransferase|Fucosylglycoprotein 3-alpha-galactosyltransferase|Fucosylglycoprotein alpha N acetylgalactosaminyltransferase|Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase|Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase soluble form|Glycoprotein fucosylgalactoside alpha galactosyltransferase|Glycoprotein fucosylgalactoside alpha N acetylgalactosaminyltransferase|Glycoprotein-fucosylgalactoside alpha-galactosyltransferase|Glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase|GTB|Histo blood group A transferase|Histo blood group A2 transferase|Histo blood group ABO system transferase|Histo blood group B transferase|Histo-blood group A transferase|Histo-blood group B transferase|NAGAT
- A transferase|A3GALNT|A3GALT1|ABO A3 transferase|Abo|ABO blood group (transferase A alpha 1-3 N acetylgalactosaminyltransferase transferase B alpha 1 3 galactosyltransferase)|ABO glycosyltransferase|ABO weak transfer|B (A) alpha 1|3 glactosyltransferase|B transferase|BGAT_HUMAN|Fucosylglycoprotein 3 alpha galactosyltransferase|Fucosylglycoprotein 3-alpha-galactosyltransferase|Fucosylglycoprotein alpha N acetylgalactosaminyltransferase|Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase|Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase soluble form|Glycoprotein fucosylgalactoside alpha galactosyltransferase|Glycoprotein fucosylgalactoside alpha N acetylgalactosaminyltransferase|Glycoprotein-fucosylgalactoside alpha-galactosyltransferase|Glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase|GTB|Histo blood group A transferase|Histo blood group A2 transferase|Histo blood group ABO system transferase|Histo blood group B transferase|Histo-blood group A transferase|Histo-blood group B transferase|NAGAT
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P16442
- P16442
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together