Anti-AP1M1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77835-936
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:AP-1 complex subunit mu-1 protein (1-160AA)
- Antigen symbol:AP1M1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q9BXS5
- Antigen synonyms:Adaptor protein complex AP-1 mu-1 subunit|Adaptor-related protein complex 1 mu-1 subunit|AP-1 complex subunit mu-1|AP-mu chain family member mu1A|Ap1m1|AP1M1_HUMAN|AP47|CLAPM2|Clathrin adaptor protein AP47|Clathrin assembly protein complex 1 medium chain 1|Clathrin assembly protein complex 1 medium chain|Clathrin assembly protein complex AP1 mu subunit|Clathrin coat assembly protein AP47|Clathrin coat-associated protein AP47|CLTNM|Golgi adaptor AP 1 47 kDa protein|Golgi adaptor HA1/AP1 adaptin mu-1 subunit|HA1 47 kDa subunit|MU 1A|Mu-adaptin 1|Mu1A-adaptin
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: AP1M1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human