Anti-SLITRK6 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: SLITRK6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77837-199
- 77837-198
- Antigen symbol:
- SLITRK6
- SLITRK6
- Antigen name:
- SLIT and NTRK-like protein 6 protein (621-821AA)
- SLIT and NTRK-like protein 6 protein (621-821AA)
- Antigen synonyms:
- 4832410J21Rik|DFNMYP|MGC119595|MGC119596|MGC119597|OTTHUMP00000066012|SLIK6_HUMAN|SLIT and NTRK like family member 6|SLIT and NTRK like protein 6|SLIT and NTRK-like protein 6|Slit and trk like 6|Slit and trk like gene 6|SLITRK 6|Slitrk6
- 4832410J21Rik|DFNMYP|MGC119595|MGC119596|MGC119597|OTTHUMP00000066012|SLIK6_HUMAN|SLIT and NTRK like family member 6|SLIT and NTRK like protein 6|SLIT and NTRK-like protein 6|Slit and trk like 6|Slit and trk like gene 6|SLITRK 6|Slitrk6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H5Y7
- Q9H5Y7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together