Anti-RASA2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA:1:2000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: RASA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77806-012
- 77806-011
- Antigen symbol:
- RASA2
- RASA2
- Antigen name:
- Synthetic peptide of RASA2
- Synthetic peptide of RASA2
- Antigen synonyms:
- GAP1m|GTPase activating protein of RAS|Ras GTPase activating protein 2|Ras GTPase-activating protein 2|RAS p21 protein activator 2|Rasa2|RASA2_HUMAN
- GAP1m|GTPase activating protein of RAS|Ras GTPase activating protein 2|Ras GTPase-activating protein 2|RAS p21 protein activator 2|Rasa2|RASA2_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15283
- Q15283
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together