Anti-PIK3C3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PIK3C3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77833-454
- 77833-455
- Antigen symbol:
- hVps34;MGC61518;Phosphatidylinositol 3 kinase catalytic subunit type 3;Phosphatidylinositol 3 kinase class 3;Phosphatidylinositol 3 kinase p100 subunit;Phosphatidylinositol 3-kinase catalytic subunit type 3;Phosphatidylinositol 3-kinase p100 subunit;Phosphoinositide 3 kinase class 3;Phosphoinositide-3-kinase class 3;PI3 kinase type 3;PI3-kinase type 3;PI3K type 3;Pik3c3;PK3C3_HUMAN;PtdIns 3 kinase type 3;PtdIns-3-kinase type 3;Vps 34;Vps34
- hVps34;MGC61518;Phosphatidylinositol 3 kinase catalytic subunit type 3;Phosphatidylinositol 3 kinase class 3;Phosphatidylinositol 3 kinase p100 subunit;Phosphatidylinositol 3-kinase catalytic subunit type 3;Phosphatidylinositol 3-kinase p100 subunit;Phosphoinositide 3 kinase class 3;Phosphoinositide-3-kinase class 3;PI3 kinase type 3;PI3-kinase type 3;PI3K type 3;Pik3c3;PK3C3_HUMAN;PtdIns 3 kinase type 3;PtdIns-3-kinase type 3;Vps 34;Vps34
- Antigen name:
- Phosphatidylinositol 3-kinase catalytic subunit type 3 protein (700-850AA)
- Phosphatidylinositol 3-kinase catalytic subunit type 3 protein (700-850AA)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NEB9
- Q8NEB9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together