Anti-RMND1 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: RMND1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77841-034
- 77841-035
- Antigen symbol:
- RMND1
- RMND1
- Antigen name:
- Required for meiotic nuclear division protein 1 homolog protein (35-160AA)
- Required for meiotic nuclear division protein 1 homolog protein (35-160AA)
- Antigen synonyms:
- bA351K16|bA351K16.3|C6orf96|chromosome 6 open reading frame 96|FLJ20627|MGC117362|MGC149570|MGC882602|required for meiotic nuclear division 1 homolog (S. cerevisiae)|Required for meiotic nuclear division protein 1 homolog|Rmnd1|RMND1_HUMAN
- bA351K16|bA351K16.3|C6orf96|chromosome 6 open reading frame 96|FLJ20627|MGC117362|MGC149570|MGC882602|required for meiotic nuclear division 1 homolog (S. cerevisiae)|Required for meiotic nuclear division protein 1 homolog|Rmnd1|RMND1_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NWS8
- Q9NWS8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together