Anti-ppsA Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: ppsA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Escherichia coli
- Catalog No:
- 77815-931
- 77815-932
- Antigen symbol:
- ppsA
- ppsA
- Antigen name:
- Recombinant Escherichia coli Phosphoenolpyruvate synthase protein (2-792AA)
- Recombinant Escherichia coli Phosphoenolpyruvate synthase protein (2-792AA)
- Antigen synonyms:
- ppsA| pps| b1702| JW1692Phosphoenolpyruvate synthase| PEP synthase| EC 2.7.9.2| Pyruvate| water dikinase
- ppsA| pps| b1702| JW1692Phosphoenolpyruvate synthase| PEP synthase| EC 2.7.9.2| Pyruvate| water dikinase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- E. coli
- E. coli
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P23538
- P23538
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together