Anti-DDX42 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC:1:20 to 1:200, IF:1:50 to 1:500
Type:
Antigen: DDX42
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77832-429
- 77832-428
- Antigen symbol:
- DDX42ATP-dependent RNA helicase DDX42;EC 3.6.4.13;DEAD box protein 42;RNA helicase-like protein;RHELP;RNA helicase-related protein;RNAHP;SF3b DEAD box protein;Splicing factor 3B-associated 125 kDa protein;SF3b125
- DDX42ATP-dependent RNA helicase DDX42;EC 3.6.4.13;DEAD box protein 42;RNA helicase-like protein;RHELP;RNA helicase-related protein;RNAHP;SF3b DEAD box protein;Splicing factor 3B-associated 125 kDa protein;SF3b125
- Antigen name:
- ATP-dependent RNA helicase DDX42 protein (519-819AA)
- ATP-dependent RNA helicase DDX42 protein (519-819AA)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86XP3
- Q86XP3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together