Anti-MSL2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MSL2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77839-562
- 77839-563
- Antigen symbol:
- MSL2
- MSL2
- Antigen name:
- E3 ubiquitin-protein ligase MSL2 protein (341-473AA)
- E3 ubiquitin-protein ligase MSL2 protein (341-473AA)
- Antigen synonyms:
- Male specific lethal 2 homolog (Drosophila)|Male specific lethal 2 homolog 1|Male specific lethal 2 homolog|Male specific lethal 2 like 1|Male-specific lethal 2 homolog|Male-specific lethal 2-like 1|Male-specific lethal-2 homolog 1|MSL-2|MSL2|MSL2-like 1|MSL2_HUMAN|MSL2L1|Ring finger protein 184
- Male specific lethal 2 homolog (Drosophila)|Male specific lethal 2 homolog 1|Male specific lethal 2 homolog|Male specific lethal 2 like 1|Male-specific lethal 2 homolog|Male-specific lethal 2-like 1|Male-specific lethal-2 homolog 1|MSL-2|MSL2|MSL2-like 1|MSL2_HUMAN|MSL2L1|Ring finger protein 184
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HCI7
- Q9HCI7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together