Anti-SLC2A4RG Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLC2A4RG
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77799-998
- 77799-999
- Antigen symbol:
- SLC2A4RG
- SLC2A4RG
- Antigen name:
- SLC2A4 regulator protein (1-100AA)
- SLC2A4 regulator protein (1-100AA)
- Antigen synonyms:
- GEF|GLUT4 enhancer factor|HDBP 1|HDBP-1|HDBP1|Huntington disease gene regulatory region binding protein 1|Huntington disease gene regulatory region-binding protein 1|S2A4R_HUMAN|Si 1 2 19|Si 1 2|SLC2A4 regulator|SLC2A4RG|Zinc finger protein 2 5 Glut4 enhancer factor
- GEF|GLUT4 enhancer factor|HDBP 1|HDBP-1|HDBP1|Huntington disease gene regulatory region binding protein 1|Huntington disease gene regulatory region-binding protein 1|S2A4R_HUMAN|Si 1 2 19|Si 1 2|SLC2A4 regulator|SLC2A4RG|Zinc finger protein 2 5 Glut4 enhancer factor
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NR83
- Q9NR83
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together