Anti-RASEF Rabbit Polyclonal Antibody
Catalog # 77796-707
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Ras-related protein Rab-45
- Antigen symbol:RASEF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q8IZ41
- Antigen synonyms:RASEF|RAB45|Ras and EF-hand domain-containing protein|Ras-related protein Rab-45
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: RASEF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse