Anti-UBE2D2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: UBE2D2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77803-837
- 77811-837
- Antigen symbol:
- UBE2D2
- UBE2D2
- Antigen name:
- Ubiquitin-conjugating enzyme E2 D2 protein (1-147AA)
- Ubiquitin-conjugating enzyme E2 D2 protein (1-147AA)
- Antigen synonyms:
- E2(17)KB2|OTTHUMP00000223475|OTTHUMP00000223476|OTTHUMP00000223477|OTTHUMP00000223478|OTTHUMP00000224375|PUBC 1|PUBC1|UB2D2_HUMAN|UBC 4|UBC 4/5|UBC4|UBC4/5|UBC4/5 homolog yeast|UBCH 5B|UBCH5B|Ubiquitin carrier protein|Ubiquitin carrier protein D2|Ubiquitin conjugating enzyme E2 17 kDa 2|Ubiquitin conjugating enzyme E2 D2|Ubiquitin conjugating enzyme E2D 2 (homologous to yeast UBC4/5)|Ubiquitin conjugating enzyme E2D 2 (UBC4/5 homolog yeast)|Ubiquitin conjugating enzyme E2D 2|Ubiquitin protein ligase D2|Ubiquitin-conjugating enzyme E2 D2|Ubiquitin-conjugating enzyme E2(17)KB 2|Ubiquitin-conjugating enzyme E2-17 kDa 2|Ubiquitin-protein ligase D2
- E2(17)KB2|OTTHUMP00000223475|OTTHUMP00000223476|OTTHUMP00000223477|OTTHUMP00000223478|OTTHUMP00000224375|PUBC 1|PUBC1|UB2D2_HUMAN|UBC 4|UBC 4/5|UBC4|UBC4/5|UBC4/5 homolog yeast|UBCH 5B|UBCH5B|Ubiquitin carrier protein|Ubiquitin carrier protein D2|Ubiquitin conjugating enzyme E2 17 kDa 2|Ubiquitin conjugating enzyme E2 D2|Ubiquitin conjugating enzyme E2D 2 (homologous to yeast UBC4/5)|Ubiquitin conjugating enzyme E2D 2 (UBC4/5 homolog yeast)|Ubiquitin conjugating enzyme E2D 2|Ubiquitin protein ligase D2|Ubiquitin-conjugating enzyme E2 D2|Ubiquitin-conjugating enzyme E2(17)KB 2|Ubiquitin-conjugating enzyme E2-17 kDa 2|Ubiquitin-protein ligase D2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P62837
- P62837
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together