Anti-ampC Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ampC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pseudomonas aeruginosa
Specifications
- Catalog No:
- 77815-695
- 77815-696
- Antigen symbol:
- ampC
- ampC
- Antigen name:
- Recombinant Pseudomonas aeruginosa Beta-lactamase protein (27-397AA)
- Recombinant Pseudomonas aeruginosa Beta-lactamase protein (27-397AA)
- Antigen synonyms:
- ampC|PA4110Beta-lactamase|EC 3.5.2.6|Cephalosporinase
- ampC|PA4110Beta-lactamase|EC 3.5.2.6|Cephalosporinase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Pseudomonas aeruginosa
- Pseudomonas aeruginosa
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P24735
- P24735
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together