Anti-ELP2 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ELP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77829-344
- 77829-343
- Antigen symbol:
- ELP2
- ELP2
- Antigen name:
- Elongator complex protein 2 protein (323-457AA)
- Elongator complex protein 2 protein (323-457AA)
- Antigen synonyms:
- AU023723 | Elongation protein 2 homolog (S. cerevisiae)| Elongator acetyltransferase complex subunit 2| Elongator complex protein 2| elongator protein 2| ELP2| ELP2_HUMAN| Epl2| FLJ10879 | SHINC 2| SHINC-2| SHINC2| signal transducer and activator of transcription 3 interacting protein 1| signal transducer and activator of transcription interacting protein 1| STAT3 interacting protein 1| Stat3 interacting protein| STAT3-interacting protein 1| STATIP 1| STATIP1| STATIP1 protein | StIP| StIP1
- AU023723 | Elongation protein 2 homolog (S. cerevisiae)| Elongator acetyltransferase complex subunit 2| Elongator complex protein 2| elongator protein 2| ELP2| ELP2_HUMAN| Epl2| FLJ10879 | SHINC 2| SHINC-2| SHINC2| signal transducer and activator of transcription 3 interacting protein 1| signal transducer and activator of transcription interacting protein 1| STAT3 interacting protein 1| Stat3 interacting protein| STAT3-interacting protein 1| STATIP 1| STATIP1| STATIP1 protein | StIP| StIP1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6IA86
- Q6IA86
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together