Anti-SGSH Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: SGSH
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77799-421
- 77799-422
- Antigen symbol:
- SGSH
- SGSH
- Antigen name:
- N-sulphoglucosamine sulphohydrolase protein (273-502AA)
- N-sulphoglucosamine sulphohydrolase protein (273-502AA)
- Antigen synonyms:
- Heparan sulfate sulfatase|Heparan sulphate sulphatase|HSS|MPS 3A|MPS3 A|MPS3A|Mucopolysaccharidosis type IIIA|N sulfoglucosamine sulfohydrolase (sulfamidase)|N sulfoglucosamine sulfohydrolase|N-sulphoglucosamine sulphohydrolase|SFMD|SPHM_HUMAN|Sulfoglucosamine sulfamidase|Sulphamidase|Sulphoglucosamine sulphamidase
- Heparan sulfate sulfatase|Heparan sulphate sulphatase|HSS|MPS 3A|MPS3 A|MPS3A|Mucopolysaccharidosis type IIIA|N sulfoglucosamine sulfohydrolase (sulfamidase)|N sulfoglucosamine sulfohydrolase|N-sulphoglucosamine sulphohydrolase|SFMD|SPHM_HUMAN|Sulfoglucosamine sulfamidase|Sulphamidase|Sulphoglucosamine sulphamidase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P51688
- P51688
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together