Anti-UBL4A Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: UBL4A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77803-908
- 77811-908
- Antigen symbol:
- UBL4A
- UBL4A
- Antigen name:
- Ubiquitin-like protein 4A protein (1-157AA)
- Ubiquitin-like protein 4A protein (1-157AA)
- Antigen synonyms:
- DX254E|DXS254E|G6PD|GDX|OTTHUMP00000061462|Ubiquitin like 4|Ubiquitin like 4A|Ubiquitin like protein 4A|Ubiquitin like protein GDX|Ubiquitin-like protein 4A|Ubiquitin-like protein GDX|UBL 4|UBL 4A|UBL4|Ubl4a|UBL4A_HUMAN
- DX254E|DXS254E|G6PD|GDX|OTTHUMP00000061462|Ubiquitin like 4|Ubiquitin like 4A|Ubiquitin like protein 4A|Ubiquitin like protein GDX|Ubiquitin-like protein 4A|Ubiquitin-like protein GDX|UBL 4|UBL 4A|UBL4|Ubl4a|UBL4A_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P11441
- P11441
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together