Anti-PRPS1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: PRPS1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77795-591
- 77795-592
- Antigen symbol:
- PRPS1
- PRPS1
- Antigen name:
- Ribose-phosphate pyrophosphokinase 1 protein (1-318AA)
- Ribose-phosphate pyrophosphokinase 1 protein (1-318AA)
- Antigen synonyms:
- ARTS|CMTX5|Deafness 2 perceptive congenital|Deafness X linked 2 perceptive congenital|DFN2|DFNX1|EC 2.7.6.1|KIAA0967|Phosphoribosyl pyrophosphate synthase I|Phosphoribosyl pyrophosphate synthetase I|PPRibP|Prps1|PRPS1_HUMAN|PRS I|PRS-I|PRSI|Ribose phosphate pyrophosphokinase I|Ribose-phosphate pyrophosphokinase 1
- ARTS|CMTX5|Deafness 2 perceptive congenital|Deafness X linked 2 perceptive congenital|DFN2|DFNX1|EC 2.7.6.1|KIAA0967|Phosphoribosyl pyrophosphate synthase I|Phosphoribosyl pyrophosphate synthetase I|PPRibP|Prps1|PRPS1_HUMAN|PRS I|PRS-I|PRSI|Ribose phosphate pyrophosphokinase I|Ribose-phosphate pyrophosphokinase 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P60891
- P60891
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together