Anti-EHD1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77842-041
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:EH domain-containing protein 1 protein (401-498AA)
- Antigen symbol:EHD1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9H4M9
- Antigen synonyms:CDABP0131|EH domain containing 1|EH domain-containing protein 1|EHD1|EHD1_HUMAN|FLJ42622|FLJ44618|H PAST|hPAST1|PAST|PAST homolog 1|PAST1|Testilin
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: EHD1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human