Anti-PARVA Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PARVA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77837-513
- 77837-512
- Antigen symbol:
- PARVA
- PARVA
- Antigen name:
- Alpha-parvin protein (2-94AA)
- Alpha-parvin protein (2-94AA)
- Antigen synonyms:
- Actopaxin| Alpha parvin| Alpha-parvin| Calponin like integrin linked kinase binding protein| Calponin-like integrin-linked kinase-binding protein| CH ILKBP| CH-ILKBP| FLJ10793| FLJ12254| Matrix remodelling associated 2| Matrix remodelling associated protein 2| Matrix-remodeling-associated protein 2| MXRA 2| MXRA2| PARV A| Parva| PARVA_HUMAN
- Actopaxin| Alpha parvin| Alpha-parvin| Calponin like integrin linked kinase binding protein| Calponin-like integrin-linked kinase-binding protein| CH ILKBP| CH-ILKBP| FLJ10793| FLJ12254| Matrix remodelling associated 2| Matrix remodelling associated protein 2| Matrix-remodeling-associated protein 2| MXRA 2| MXRA2| PARV A| Parva| PARVA_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NVD7
- Q9NVD7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together