Anti-QPCTL Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: QPCTL
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77837-588
- 77837-587
- Antigen symbol:
- QPCTL
- QPCTL
- Antigen name:
- Glutaminyl-peptide cyclotransferase-like protein (212-382AA)
- Glutaminyl-peptide cyclotransferase-like protein (212-382AA)
- Antigen synonyms:
- QPCTLGlutaminyl-peptide cyclotransferase-like protein|EC 2.3.2.5|Golgi-resident glutaminyl-peptide cyclotransferase|isoQC|gQC
- QPCTLGlutaminyl-peptide cyclotransferase-like protein|EC 2.3.2.5|Golgi-resident glutaminyl-peptide cyclotransferase|isoQC|gQC
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NXS2
- Q9NXS2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together