Anti-HOXB4 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000
Type:
Antigen: HOXB4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77807-752
- 77807-753
- Antigen symbol:
- HOXB4
- HOXB4
- Antigen name:
- Synthetic peptide of HOXB4
- Synthetic peptide of HOXB4
- Antigen synonyms:
- Homeo box 2F|Homeo box B4|Homeobox 2F|Homeobox B4|Homeobox protein Hox B4|Homeobox protein Hox-2.6|Homeobox protein Hox-2F|Homeobox protein Hox-B4|Homeobox protein HoxB4|HOX 2|Hox 2.6|Hox 2F|HOX B4|HOX2|Hox2.6|Hox2F|HOXB 4|hoxb4|HXB4_HUMAN
- Homeo box 2F|Homeo box B4|Homeobox 2F|Homeobox B4|Homeobox protein Hox B4|Homeobox protein Hox-2.6|Homeobox protein Hox-2F|Homeobox protein Hox-B4|Homeobox protein HoxB4|HOX 2|Hox 2.6|Hox 2F|HOX B4|HOX2|Hox2.6|Hox2F|HOXB 4|hoxb4|HXB4_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P17483
- P17483
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together