Anti-TRIM26 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TRIM26
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-819
- 77823-818
- Antigen symbol:
- Acid finger protein;AFP;CAM25547;RING finger protein 95;RNF95;TRI26_HUMAN;TRIM26;tripartite motif containing 26;Tripartite motif-containing protein 26;widely expressed acid zinc finger protein;Zinc finger protein 173;formerly;ZNF173
- Acid finger protein;AFP;CAM25547;RING finger protein 95;RNF95;TRI26_HUMAN;TRIM26;tripartite motif containing 26;Tripartite motif-containing protein 26;widely expressed acid zinc finger protein;Zinc finger protein 173;formerly;ZNF173
- Antigen name:
- Tripartite motif-containing protein 26 protein (151-270AA)
- Tripartite motif-containing protein 26 protein (151-270AA)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q12899
- Q12899
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together