Anti-USH1C Rabbit Polyclonal Antibody
77804-089
New Product
- Antigen name:Usher syndrome type-1C protein
- Antigen symbol:USH1C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9Y6N9
- Antigen synonyms:AIE 75| AIE75| Antigen NY CO 38/NY CO 37| Antigen NY-CO-38/NY-CO-37| Autoimmune enteropathy related antigen AIE 75| Autoimmune enteropathy related antigen AIE75| Autoimmune enteropathy-related antigen AIE-75| Deafness autosomal recessive 18| DFNB 18| DFNB18| Harmonin| NY CO 37| NY CO 38| PDZ 45| PDZ 73| PDZ 73 protein| PDZ 73/NY CO 38| PDZ45| PDZ73| PDZ73 protein| Protein PDZ-73| Renal carcinoma antigen NY REN 3| Renal carcinoma antigen NY-REN-3| USH 1C| USH1C| USH1C_HUMAN| Ush1cpst| Usher syndrome 1C (autosomal recessive severe)| Usher syndrome 1C| Usher syndrome type 1C protein| Usher syndrome type-1C protein
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: USH1C
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat