Anti-TMEM209 Rabbit Recombinant Polyclonal Antibody (Biotin)
New Product
Type:
Antigen: TMEM209
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77811-025
- 77803-025
- Antigen symbol:
- TMEM209
- TMEM209
- Antigen name:
- Transmembrane protein 209 protein (251-519AA)
- Transmembrane protein 209 protein (251-519AA)
- Antigen synonyms:
- TMEM209|Transmembrane protein 209
- TMEM209|Transmembrane protein 209
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96SK2
- Q96SK2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together