Anti-NMRAL1 Rabbit Recombinant Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NMRAL1
Clonality: Recombinant
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77837-295
- 77837-296
- Antigen symbol:
- NMRAL1
- NMRAL1
- Antigen name:
- NmrA-like family domain-containing protein 1 protein (1-299AA)
- NmrA-like family domain-containing protein 1 protein (1-299AA)
- Antigen synonyms:
- FLJ25918|HSCARG|NmrA like family domain containing 1|NmrA like family domain containing protein 1|NmrA-like family domain-containing protein 1|NMRAL1|NMRL1_HUMAN|SDR48A1
- FLJ25918|HSCARG|NmrA like family domain containing 1|NmrA like family domain containing protein 1|NmrA-like family domain-containing protein 1|NMRAL1|NMRL1_HUMAN|SDR48A1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HBL8
- Q9HBL8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together