Order Entry
Canada
ContactUsLinkComponent
Anti-SPIN2A Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Anti-SPIN2A Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-SPIN2A Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77834-877
  • 77834-878
  • Antigen symbol:
  • SPIN2A
  • SPIN2A
  • Antigen name:
  • Spindlin-2A protein (1-258AA)
  • Spindlin-2A protein (1-258AA)
  • Antigen synonyms:
  • SPIN2A|DXF34|SPIN2|Spindlin-2A|Protein DXF34|Spindlin-like protein 2A|SPIN-2|SPIN-2A
  • SPIN2A|DXF34|SPIN2|Spindlin-2A|Protein DXF34|Spindlin-like protein 2A|SPIN-2|SPIN-2A
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q99865
  • Q99865
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together