Anti-LPAL2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LPAL2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-277
- 77823-276
- Antigen symbol:
- LPAL2
- LPAL2
- Antigen name:
- Putative apolipoprotein(a)-like protein 2 protein (52-124AA)
- Putative apolipoprotein(a)-like protein 2 protein (52-124AA)
- Antigen synonyms:
- LPAL2|APOARGC|Putative apolipoprotein(a)-like protein 2|Apo(a)-like protein 2|Lp(a)-liker protein 2|Apolipoprotein a-related gene C protein|Apo(a)rg-C
- LPAL2|APOARGC|Putative apolipoprotein(a)-like protein 2|Apo(a)-like protein 2|Lp(a)-liker protein 2|Apolipoprotein a-related gene C protein|Apo(a)rg-C
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q16609
- Q16609
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together