Anti-NBPF12/NBPF10/NBPF1/NBPF9 Rabbit Polyclonal Antibody
Catalog # 77813-614
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Synthesized peptide derived from internal of NBPF1/9/10/12/14/15/16/20.
- Antigen symbol:NBPF12/NBPF10/NBPF1/NBPF9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µl
- Form:liquid
- Gene ID:Q5TAG4
- Antigen synonyms:NBPF12|COAS1|KIAA1245|Neuroblastoma breakpoint family member 12|Chromosome 1 amplified sequence 1
- Storage buffer:PBS, 150mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: NBPF12/NBPF10/NBPF1/NBPF9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human