Anti-MSA2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MSA2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plasmodium falciparum
Specifications
- Catalog No:
- 77816-430
- 77816-431
- Antigen symbol:
- MSA2
- MSA2
- Antigen name:
- Recombinant Plasmodium falciparum Merozoite surface antigen 2 protein (109-246AA)
- Recombinant Plasmodium falciparum Merozoite surface antigen 2 protein (109-246AA)
- Antigen synonyms:
- MSA2|PFB0300c|Merozoite surface antigen 2|MSA-2|45 kDa merozoite surface antigen
- MSA2|PFB0300c|Merozoite surface antigen 2|MSA-2|45 kDa merozoite surface antigen
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Plasmodium falciparum
- Plasmodium falciparum
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P50498
- P50498
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together