Anti-TREH Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TREH
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-497
- 77803-497
- Antigen symbol:
- TREH
- TREH
- Antigen name:
- Trehalase protein (373-507AA)
- Trehalase protein (373-507AA)
- Antigen synonyms:
- Alpha alpha trehalase|Alpha alpha trehalose glucohydrolase|Alpha|alpha-trehalase|alpha-trehalose glucohydrolase|Brush border membrane glycoprotein|MGC129621|TRE|TREA|TREA_HUMAN|Trehalase
- Alpha alpha trehalase|Alpha alpha trehalose glucohydrolase|Alpha|alpha-trehalase|alpha-trehalose glucohydrolase|Brush border membrane glycoprotein|MGC129621|TRE|TREA|TREA_HUMAN|Trehalase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43280
- O43280
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together