Anti-TAC1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TAC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77801-773
- 77801-774
- Antigen symbol:
- TAC1
- TAC1
- Antigen name:
- Protachykinin-1 protein (1-129AA)
- Protachykinin-1 protein (1-129AA)
- Antigen synonyms:
- C-terminal-flanking peptide|Hs.2563|neurokinin 1|neurokinin 2|neurokinin A|neurokinin alpha|Neuromedin L|neuropeptide gamma|neuropeptide K|NK2|NKA|NKNA|NPK|PPT|preprotachykinin|protachykinin|protachykinin-1|Substance K|SubstanceP|TAC1|TAC2|formerly|Tachykinin 1|tachykinin 2|Tachykinin precursor 1|tachykinin|precursor 1 (substance K|substance P|neuromedin L|neuropeptide gamma)|Tachykinin1|TKN1_HUMAN
- C-terminal-flanking peptide|Hs.2563|neurokinin 1|neurokinin 2|neurokinin A|neurokinin alpha|Neuromedin L|neuropeptide gamma|neuropeptide K|NK2|NKA|NKNA|NPK|PPT|preprotachykinin|protachykinin|protachykinin-1|Substance K|SubstanceP|TAC1|TAC2|formerly|Tachykinin 1|tachykinin 2|Tachykinin precursor 1|tachykinin|precursor 1 (substance K|substance P|neuromedin L|neuropeptide gamma)|Tachykinin1|TKN1_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P20366
- P20366
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together