Anti-RBMY1A1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000
Type:
Antigen: RBMY1A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77821-042
- 77821-041
- Antigen symbol:
- RBMY1A1
- RBMY1A1
- Antigen name:
- Fusion protein of RBMY1A1
- Fusion protein of RBMY1A1
- Antigen synonyms:
- family 1 member A1|hRBMY|MGC181956|RBM|RBM1|RBM2|RBMY|RBMY1C|RBY1A_HUMAN|RNA binding motif protein 1|RNA binding motif protein 2|RNA binding motif protein Y chromosome family 1 member A1|RNA binding motif protein Y chromosome family 1 member A1/C|RNA binding motif protein Y chromosome family 1 member|RNA binding motif protein Y linked family 1 member A1|RNA-binding motif protein 1|RNA-binding motif protein 2|RNA-binding motif protein|Y chromosome|Y chromosome RNA recognition motif 1|YRRM1|YRRM2
- family 1 member A1|hRBMY|MGC181956|RBM|RBM1|RBM2|RBMY|RBMY1C|RBY1A_HUMAN|RNA binding motif protein 1|RNA binding motif protein 2|RNA binding motif protein Y chromosome family 1 member A1|RNA binding motif protein Y chromosome family 1 member A1/C|RNA binding motif protein Y chromosome family 1 member|RNA binding motif protein Y linked family 1 member A1|RNA-binding motif protein 1|RNA-binding motif protein 2|RNA-binding motif protein|Y chromosome|Y chromosome RNA recognition motif 1|YRRM1|YRRM2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P0DJD3
- P0DJD3
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together